Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC65
Entrez Gene ID
85478
Gene Name
coiled-coil domain containing 65
Gene Aliases
CFAP250, CILD27, DRC2, FAP250, NYD-SP28
Chromosome Location
Chr.12: 48904110 - 48921576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286957.1 NP_001273886.1 3 - 4 661 94
NM_033124.4 NP_149115.2 3 - 4 701 94
GenBank mRNA AF382188.1 - 3 - 4 666 94
AK093051.1 - 3 - 4 674 94
AK302082.1 - 3 - 4 638 94
AK314623.1 - 3 - 4 646 94
BC039317.1 - 3 - 4 640 94

View Full Product Details