The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145845.1 | NP_001139317.1 | 8 - 9 | 2056 | 82 |
NM_002941.3 | NP_002932.1 | 9 - 10 | 1374 | 82 | |
NM_133631.3 | NP_598334.2 | 8 - 9 | 2056 | 82 | |
XM_006713277.2 | XP_006713340.1 | 8 - 9 | 2025 | 82 | |
XM_011533976.1 | XP_011532278.1 | 10 - 11 | 1472 | 82 | |
XM_011533977.2 | XP_011532279.1 | 10 - 11 | 1325 | 82 | |
XM_011533978.1 | XP_011532280.1 | 10 - 11 | 1472 | 82 | |
XM_011533979.1 | XP_011532281.1 | 10 - 11 | 1472 | 82 | |
XM_011533980.1 | XP_011532282.1 | 10 - 11 | 1472 | 82 | |
XM_017006982.1 | XP_016862471.1 | 9 - 10 | 1384 | 82 | |
XM_017006983.1 | XP_016862472.1 | 7 - 8 | 2016 | 82 | |
XM_017006984.1 | XP_016862473.1 | 9 - 10 | 1463 | 82 | |
XM_017006985.1 | XP_016862474.1 | 8 - 9 | 1125 | 82 | |
GenBank mRNA | AF040990.1 | - | 8 - 9 | 1166 | 82 |
AK095256.1 | - | 3 - 4 | 991 | 82 | |
BC112336.1 | - | 8 - 9 | 1109 | 82 | |
BC115020.1 | - | 8 - 9 | 1157 | 82 | |
BC115022.1 | - | 8 - 9 | 1157 | 82 | |
BC157861.1 | - | 8 - 9 | 1353 | 82 | |
BC171855.1 | - | 8 - 9 | 1353 | 82 | |
BX538319.1 | - | 1 - 2 | 73 | 82 |