The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145845.1 | NP_001139317.1 | 10 - 11 | 2434 | 65 |
NM_002941.3 | NP_002932.1 | 11 - 12 | 1752 | 65 | |
NM_133631.3 | NP_598334.2 | 10 - 11 | 2434 | 65 | |
XM_006713277.2 | XP_006713340.1 | 10 - 11 | 2403 | 65 | |
XM_011533976.1 | XP_011532278.1 | 12 - 13 | 1850 | 65 | |
XM_011533977.2 | XP_011532279.1 | 12 - 13 | 1703 | 65 | |
XM_011533978.1 | XP_011532280.1 | 12 - 13 | 1850 | 65 | |
XM_011533979.1 | XP_011532281.1 | 12 - 13 | 1850 | 65 | |
XM_011533980.1 | XP_011532282.1 | 12 - 13 | 1850 | 65 | |
XM_017006982.1 | XP_016862471.1 | 11 - 12 | 1762 | 65 | |
XM_017006983.1 | XP_016862472.1 | 9 - 10 | 2394 | 65 | |
XM_017006984.1 | XP_016862473.1 | 11 - 12 | 1841 | 65 | |
XM_017006985.1 | XP_016862474.1 | 10 - 11 | 1503 | 65 | |
GenBank mRNA | AF040990.1 | - | 10 - 11 | 1544 | 65 |
AK095256.1 | - | 5 - 6 | 1369 | 65 | |
AK299920.1 | - | 1 - 2 | 441 | 65 | |
BC112336.1 | - | 10 - 11 | 1487 | 65 | |
BC115020.1 | - | 10 - 11 | 1535 | 65 | |
BC115022.1 | - | 10 - 11 | 1535 | 65 | |
BC157861.1 | - | 10 - 11 | 1731 | 65 | |
BC171855.1 | - | 10 - 11 | 1731 | 65 | |
BX538319.1 | - | 3 - 4 | 451 | 65 |