The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145845.1 | NP_001139317.1 | 14 - 15 | 2975 | 94 |
NM_002941.3 | NP_002932.1 | 15 - 16 | 2293 | 94 | |
NM_133631.3 | NP_598334.2 | 14 - 15 | 2975 | 94 | |
XM_006713277.2 | XP_006713340.1 | 14 - 15 | 2944 | 94 | |
XM_011533976.1 | XP_011532278.1 | 16 - 17 | 2391 | 94 | |
XM_011533977.2 | XP_011532279.1 | 16 - 17 | 2244 | 94 | |
XM_011533978.1 | XP_011532280.1 | 16 - 17 | 2391 | 94 | |
XM_011533979.1 | XP_011532281.1 | 16 - 17 | 2391 | 94 | |
XM_011533980.1 | XP_011532282.1 | 16 - 17 | 2391 | 94 | |
XM_017006982.1 | XP_016862471.1 | 15 - 16 | 2303 | 94 | |
XM_017006983.1 | XP_016862472.1 | 13 - 14 | 2935 | 94 | |
XM_017006984.1 | XP_016862473.1 | 15 - 16 | 2382 | 94 | |
XM_017006985.1 | XP_016862474.1 | 14 - 15 | 2044 | 94 | |
GenBank mRNA | AF040990.1 | - | 14 - 15 | 2085 | 94 |
AK095256.1 | - | 9 - 10 | 1910 | 94 | |
AK299920.1 | - | 5 - 6 | 982 | 94 | |
BC112336.1 | - | 14 - 15 | 2028 | 94 | |
BC115022.1 | - | 14 - 15 | 2076 | 94 | |
BC157861.1 | - | 14 - 15 | 2272 | 94 | |
BC171855.1 | - | 14 - 15 | 2272 | 94 | |
BX538319.1 | - | 7 - 8 | 1004 | 94 |