The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145845.1 | NP_001139317.1 | 15 - 16 | 3211 | 101 |
NM_002941.3 | NP_002932.1 | 16 - 17 | 2529 | 101 | |
NM_133631.3 | NP_598334.2 | 15 - 16 | 3211 | 101 | |
XM_006713277.2 | XP_006713340.1 | 15 - 16 | 3180 | 101 | |
XM_011533976.1 | XP_011532278.1 | 17 - 18 | 2627 | 101 | |
XM_011533977.2 | XP_011532279.1 | 17 - 18 | 2480 | 101 | |
XM_011533978.1 | XP_011532280.1 | 17 - 18 | 2627 | 101 | |
XM_011533979.1 | XP_011532281.1 | 17 - 18 | 2627 | 101 | |
XM_011533980.1 | XP_011532282.1 | 17 - 18 | 2627 | 101 | |
XM_017006982.1 | XP_016862471.1 | 16 - 17 | 2539 | 101 | |
XM_017006983.1 | XP_016862472.1 | 14 - 15 | 3171 | 101 | |
XM_017006984.1 | XP_016862473.1 | 16 - 17 | 2618 | 101 | |
XM_017006985.1 | XP_016862474.1 | 15 - 16 | 2280 | 101 | |
GenBank mRNA | AF040990.1 | - | 15 - 16 | 2321 | 101 |
AK299920.1 | - | 6 - 7 | 1218 | 101 | |
BC112336.1 | - | 15 - 16 | 2264 | 101 | |
BC115022.1 | - | 15 - 16 | 2312 | 101 | |
BC157861.1 | - | 15 - 16 | 2508 | 101 | |
BC171855.1 | - | 15 - 16 | 2508 | 101 | |
BX538319.1 | - | 8 - 9 | 1240 | 101 |