The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145845.1 | NP_001139317.1 | 17 - 18 | 3508 | 59 |
NM_002941.3 | NP_002932.1 | 18 - 19 | 2826 | 59 | |
NM_133631.3 | NP_598334.2 | 17 - 18 | 3508 | 59 | |
XM_006713277.2 | XP_006713340.1 | 17 - 18 | 3477 | 59 | |
XM_011533976.1 | XP_011532278.1 | 19 - 20 | 2924 | 59 | |
XM_011533977.2 | XP_011532279.1 | 19 - 20 | 2777 | 59 | |
XM_011533978.1 | XP_011532280.1 | 19 - 20 | 2924 | 59 | |
XM_011533979.1 | XP_011532281.1 | 19 - 20 | 2924 | 59 | |
XM_011533980.1 | XP_011532282.1 | 19 - 20 | 2924 | 59 | |
XM_017006982.1 | XP_016862471.1 | 18 - 19 | 2836 | 59 | |
XM_017006983.1 | XP_016862472.1 | 16 - 17 | 3468 | 59 | |
XM_017006984.1 | XP_016862473.1 | 18 - 19 | 2915 | 59 | |
XM_017006985.1 | XP_016862474.1 | 17 - 18 | 2577 | 59 | |
GenBank mRNA | AF040990.1 | - | 17 - 18 | 2618 | 59 |
AK299920.1 | - | 8 - 9 | 1515 | 59 | |
BC112336.1 | - | 17 - 18 | 2561 | 59 | |
BC115020.1 | - | 17 - 18 | 2609 | 59 | |
BC115022.1 | - | 17 - 18 | 2609 | 59 | |
BC157861.1 | - | 17 - 18 | 2805 | 59 | |
BC171855.1 | - | 17 - 18 | 2805 | 59 | |
BX538319.1 | - | 10 - 11 | 1537 | 59 |