The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002941.3 | NP_002932.1 | 20 - 21 | 3046 | 69 |
XM_006713277.2 | XP_006713340.1 | 19 - 20 | 3697 | 69 | |
XM_011533976.1 | XP_011532278.1 | 21 - 22 | 3144 | 69 | |
XM_011533977.2 | XP_011532279.1 | 21 - 22 | 2997 | 69 | |
XM_011533979.1 | XP_011532281.1 | 21 - 22 | 3144 | 69 | |
XM_017006982.1 | XP_016862471.1 | 20 - 21 | 3056 | 69 | |
XM_017006983.1 | XP_016862472.1 | 18 - 19 | 3688 | 69 | |
XM_017006984.1 | XP_016862473.1 | 20 - 21 | 3135 | 69 | |
XM_017006985.1 | XP_016862474.1 | 19 - 20 | 2797 | 69 | |
GenBank mRNA | AF040990.1 | - | 19 - 20 | 2838 | 69 |
AK299920.1 | - | 10 - 11 | 1735 | 69 | |
BX538319.1 | - | 12 - 13 | 1757 | 69 |