The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145845.1 | NP_001139317.1 | 19 - 20 | 3749 | 63 |
NM_002941.3 | NP_002932.1 | 21 - 22 | 3094 | 63 | |
NM_133631.3 | NP_598334.2 | 19 - 20 | 3749 | 63 | |
XM_006713277.2 | XP_006713340.1 | 20 - 21 | 3745 | 63 | |
XM_011533976.1 | XP_011532278.1 | 22 - 23 | 3192 | 63 | |
XM_011533977.2 | XP_011532279.1 | 22 - 23 | 3045 | 63 | |
XM_011533978.1 | XP_011532280.1 | 21 - 22 | 3165 | 63 | |
XM_011533979.1 | XP_011532281.1 | 22 - 23 | 3192 | 63 | |
XM_011533980.1 | XP_011532282.1 | 21 - 22 | 3165 | 63 | |
XM_017006982.1 | XP_016862471.1 | 21 - 22 | 3104 | 63 | |
XM_017006983.1 | XP_016862472.1 | 19 - 20 | 3736 | 63 | |
XM_017006984.1 | XP_016862473.1 | 21 - 22 | 3183 | 63 | |
XM_017006985.1 | XP_016862474.1 | 20 - 21 | 2845 | 63 | |
GenBank mRNA | AF040990.1 | - | 20 - 21 | 2886 | 63 |
AK299920.1 | - | 11 - 12 | 1783 | 63 | |
BC112336.1 | - | 19 - 20 | 2802 | 63 | |
BC115020.1 | - | 19 - 20 | 2850 | 63 | |
BC115022.1 | - | 19 - 20 | 2850 | 63 | |
BC157861.1 | - | 19 - 20 | 3046 | 63 | |
BC171855.1 | - | 19 - 20 | 3046 | 63 | |
BX538319.1 | - | 13 - 14 | 1805 | 63 |