The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002941.3 | NP_002932.1 | 22 - 23 | 3247 | 92 |
NM_133631.3 | NP_598334.2 | 20 - 21 | 3902 | 92 | |
XM_006713277.2 | XP_006713340.1 | 21 - 22 | 3898 | 92 | |
XM_011533976.1 | XP_011532278.1 | 23 - 24 | 3345 | 92 | |
XM_011533977.2 | XP_011532279.1 | 23 - 24 | 3198 | 92 | |
XM_011533978.1 | XP_011532280.1 | 22 - 23 | 3318 | 92 | |
XM_017006982.1 | XP_016862471.1 | 22 - 23 | 3257 | 92 | |
XM_017006983.1 | XP_016862472.1 | 20 - 21 | 3889 | 92 | |
XM_017006985.1 | XP_016862474.1 | 21 - 22 | 2998 | 92 | |
GenBank mRNA | AF040990.1 | - | 21 - 22 | 3039 | 92 |
AK299920.1 | - | 12 - 13 | 1936 | 92 | |
BC112336.1 | - | 20 - 21 | 2955 | 92 | |
BC115020.1 | - | 20 - 21 | 3003 | 92 | |
BC157861.1 | - | 20 - 21 | 3199 | 92 | |
BC171855.1 | - | 20 - 21 | 3199 | 92 | |
BX538319.1 | - | 14 - 15 | 1958 | 92 |