The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145845.1 | NP_001139317.1 | 24 - 25 | 4580 | 66 |
NM_002941.3 | NP_002932.1 | 26 - 27 | 4090 | 66 | |
NM_133631.3 | NP_598334.2 | 24 - 25 | 4745 | 66 | |
XM_006713277.2 | XP_006713340.1 | 25 - 26 | 4741 | 66 | |
XM_011533976.1 | XP_011532278.1 | 27 - 28 | 4188 | 66 | |
XM_011533977.2 | XP_011532279.1 | 27 - 28 | 4041 | 66 | |
XM_011533978.1 | XP_011532280.1 | 26 - 27 | 4161 | 66 | |
XM_011533979.1 | XP_011532281.1 | 27 - 28 | 4023 | 66 | |
XM_011533980.1 | XP_011532282.1 | 26 - 27 | 3996 | 66 | |
XM_017006982.1 | XP_016862471.1 | 26 - 27 | 4100 | 66 | |
XM_017006983.1 | XP_016862472.1 | 24 - 25 | 4732 | 66 | |
XM_017006984.1 | XP_016862473.1 | 26 - 27 | 4014 | 66 | |
XM_017006985.1 | XP_016862474.1 | 25 - 26 | 3841 | 66 | |
GenBank mRNA | AF040990.1 | - | 25 - 26 | 3882 | 66 |
AK299920.1 | - | 16 - 17 | 2779 | 66 | |
BC001969.1 | - | 2 - 3 | 413 | 66 | |
BC112336.1 | - | 24 - 25 | 3801 | 66 | |
BC115020.1 | - | 24 - 25 | 3846 | 66 | |
BC115022.1 | - | 24 - 25 | 3681 | 66 | |
BC157861.1 | - | 24 - 25 | 4042 | 66 | |
BC171855.1 | - | 24 - 25 | 4042 | 66 | |
BX538319.1 | - | 18 - 19 | 2801 | 66 |