The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145845.1 | NP_001139317.1 | 4 - 5 | 1656 | 123 |
NM_002941.3 | NP_002932.1 | 6 - 7 | 983 | 123 | |
NM_133631.3 | NP_598334.2 | 4 - 5 | 1656 | 123 | |
XM_006713277.2 | XP_006713340.1 | 4 - 5 | 1625 | 123 | |
XM_011533976.1 | XP_011532278.1 | 6 - 7 | 1072 | 123 | |
XM_011533977.2 | XP_011532279.1 | 6 - 7 | 925 | 123 | |
XM_011533978.1 | XP_011532280.1 | 6 - 7 | 1072 | 123 | |
XM_011533979.1 | XP_011532281.1 | 6 - 7 | 1072 | 123 | |
XM_011533980.1 | XP_011532282.1 | 6 - 7 | 1072 | 123 | |
XM_017006982.1 | XP_016862471.1 | 5 - 6 | 984 | 123 | |
XM_017006983.1 | XP_016862472.1 | 4 - 5 | 1625 | 123 | |
XM_017006984.1 | XP_016862473.1 | 6 - 7 | 1072 | 123 | |
XM_017006985.1 | XP_016862474.1 | 4 - 5 | 725 | 123 | |
GenBank mRNA | AF040990.1 | - | 5 - 6 | 775 | 123 |
BC112336.1 | - | 4 - 5 | 709 | 123 | |
BC115022.1 | - | 4 - 5 | 757 | 123 | |
BC157861.1 | - | 4 - 5 | 953 | 123 | |
BC171855.1 | - | 4 - 5 | 953 | 123 |