Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIP2B
Entrez Gene ID
57609
Gene Name
disco interacting protein 2 homolog B
Gene Aliases
-
Chromosome Location
Chr.12: 50504985 - 50748667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173602.2 NP_775873.2 2 - 3 331 108
XM_005269044.1 XP_005269101.1 2 - 3 331 108
XM_006719520.3 XP_006719583.1 2 - 3 277 108
XM_006719521.3 XP_006719584.1 2 - 3 327 108
XM_011538594.2 XP_011536896.1 2 - 3 327 108
XM_011538595.2 XP_011536897.1 2 - 3 327 108
XM_011538596.2 XP_011536898.1 2 - 3 327 108
XM_017019699.1 XP_016875188.1 2 - 3 327 108
XM_017019700.1 XP_016875189.1 2 - 3 331 108
XM_017019701.1 XP_016875190.1 2 - 3 331 108
XM_017019702.1 XP_016875191.1 2 - 3 331 108
XM_017019703.1 XP_016875192.1 2 - 3 320 108
XM_017019704.1 XP_016875193.1 2 - 3 331 108
XM_017019705.1 XP_016875194.1 2 - 3 331 108
GenBank mRNA AK091597.1 - 2 - 3 331 108
BC075027.2 - 2 - 3 209 108

View Full Product Details