Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIP2B
Entrez Gene ID
57609
Gene Name
disco interacting protein 2 homolog B
Gene Aliases
-
Chromosome Location
Chr.12: 50504985 - 50748667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173602.2 NP_775873.2 36 - 37 4507 60
XM_005269044.1 XP_005269101.1 36 - 37 4510 60
XM_006719520.3 XP_006719583.1 37 - 38 4483 60
XM_006719521.3 XP_006719584.1 36 - 37 4320 60
XM_011538594.2 XP_011536896.1 37 - 38 4536 60
XM_011538595.2 XP_011536897.1 35 - 36 4368 60
XM_011538596.2 XP_011536898.1 36 - 37 4323 60
XM_017019699.1 XP_016875188.1 37 - 38 4518 60
XM_017019700.1 XP_016875189.1 36 - 37 4492 60
XM_017019701.1 XP_016875190.1 36 - 37 4489 60
XM_017019702.1 XP_016875191.1 34 - 35 4339 60
XM_017019703.1 XP_016875192.1 35 - 36 4286 60
XM_017019704.1 XP_016875193.1 35 - 36 4294 60
XM_017019705.1 XP_016875194.1 35 - 36 4276 60
GenBank mRNA AB040896.2 - 27 - 28 3121 60
AK097369.1 - 18 - 19 2063 60
AK126758.1 - 25 - 26 2878 60
BC007671.2 - 20 - 21 2431 60
BC030156.2 - 20 - 21 2431 60

View Full Product Details