Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFAP2B
Entrez Gene ID
7021
Gene Name
transcription factor AP-2 beta
Gene Aliases
AP-2B, AP2-B
Chromosome Location
Chr.6: 50817692 - 50847619 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003221.3 NP_003212.2 3 - 4 768 93
XM_011514837.2 XP_011513139.1 4 - 5 1073 93
XM_017011233.1 XP_016866722.1 6 - 7 1449 93
XM_017011234.1 XP_016866723.1 6 - 7 902 93
XM_017011235.1 XP_016866724.1 2 - 3 581 93
GenBank mRNA AK291172.1 - 3 - 4 608 93
AK313749.1 - 3 - 4 606 93
BC037225.1 - 3 - 4 624 93
X95694.1 - 3 - 4 601 93

View Full Product Details