The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003221.3 | NP_003212.2 | 5 - 6 | 1108 | 79 |
XM_011514837.2 | XP_011513139.1 | 6 - 7 | 1413 | 79 | |
XM_017011233.1 | XP_016866722.1 | 8 - 9 | 1789 | 79 | |
XM_017011234.1 | XP_016866723.1 | 8 - 9 | 1242 | 79 | |
XM_017011235.1 | XP_016866724.1 | 4 - 5 | 921 | 79 | |
GenBank mRNA | AK291172.1 | - | 5 - 6 | 948 | 79 |
AK313749.1 | - | 5 - 6 | 946 | 79 | |
BC037225.1 | - | 5 - 6 | 964 | 79 | |
X95694.1 | - | 5 - 6 | 941 | 79 |