The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003221.3 | NP_003212.2 | 1 - 2 | 249 | 121 |
GenBank mRNA | AK291172.1 | - | 1 - 2 | 89 | 121 |
AK313749.1 | - | 1 - 2 | 87 | 121 | |
AU141084.1 | - | 1 - 2 | 249 | 121 | |
BC037225.1 | - | 1 - 2 | 105 | 121 | |
X95694.1 | - | 1 - 2 | 82 | 121 |