The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003221.3 | NP_003212.2 | 6 - 7 | 1250 | 67 |
XM_011514837.2 | XP_011513139.1 | 7 - 8 | 1555 | 67 | |
XM_017011233.1 | XP_016866722.1 | 9 - 10 | 1931 | 67 | |
XM_017011234.1 | XP_016866723.1 | 9 - 10 | 1384 | 67 | |
XM_017011235.1 | XP_016866724.1 | 5 - 6 | 1063 | 67 | |
GenBank mRNA | AK291172.1 | - | 6 - 7 | 1090 | 67 |
AK313749.1 | - | 6 - 7 | 1088 | 67 | |
BC037225.1 | - | 6 - 7 | 1106 | 67 | |
X95694.1 | - | 6 - 7 | 1083 | 67 |