The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009998.3 | NP_001009998.1 | 11 - 12 | 1030 | 85 |
NM_032627.4 | NP_116016.1 | 12 - 13 | 1096 | 85 | |
XM_005259790.3 | XP_005259847.1 | 12 - 13 | 1137 | 85 | |
XM_006722665.3 | XP_006722728.1 | 12 - 13 | 1143 | 85 | |
XM_006722666.2 | XP_006722729.1 | 12 - 13 | 1101 | 85 | |
XM_006722668.2 | XP_006722731.1 | 11 - 12 | 1035 | 85 | |
XM_017026431.1 | XP_016881920.1 | 12 - 13 | 1155 | 85 | |
XM_017026432.1 | XP_016881921.1 | 12 - 13 | 1693 | 85 | |
XM_017026433.1 | XP_016881922.1 | 12 - 13 | 1101 | 85 | |
XM_017026434.1 | XP_016881923.1 | 11 - 12 | 1084 | 85 | |
XM_017026435.1 | XP_016881924.1 | 11 - 12 | 1626 | 85 | |
XM_017026436.1 | XP_016881925.1 | 11 - 12 | 1035 | 85 | |
XM_017026437.1 | XP_016881926.1 | 11 - 12 | 1066 | 85 | |
GenBank mRNA | BC000274.1 | - | 12 - 13 | 1003 | 85 |