The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242614.1 | NP_001229543.1 | 5 - 6 | 582 | 72 |
NM_031462.3 | NP_113650.2 | 5 - 6 | 570 | 72 | |
NM_134446.3 | NP_604395.1 | 3 - 4 | 423 | 72 | |
XM_017029890.1 | XP_016885379.1 | 5 - 6 | 434 | 72 | |
XM_017029891.1 | XP_016885380.1 | 3 - 4 | 421 | 72 | |
GenBank mRNA | AK024278.1 | - | 1 - 2 | 77 | 72 |
AK125020.1 | - | 5 - 6 | 393 | 72 | |
AK290200.1 | - | 5 - 6 | 346 | 72 | |
AK291375.1 | - | 3 - 4 | 236 | 72 | |
AK295217.1 | - | 5 - 6 | 388 | 72 | |
AL136580.1 | - | 5 - 6 | 460 | 72 | |
AY078165.1 | - | 5 - 6 | 378 | 72 | |
AY078166.1 | - | 3 - 4 | 231 | 72 | |
AY358837.1 | - | 5 - 6 | 443 | 72 | |
BC007569.1 | - | 4 - 5 | 230 | 72 | |
BC013029.2 | - | 4 - 5 | 230 | 72 | |
BC025729.1 | - | 6 - 7 | 1683 | 72 | |
BC030536.1 | - | 5 - 6 | 430 | 72 | |
KU178797.1 | - | 5 - 6 | 342 | 72 |