Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD99L2
Entrez Gene ID
83692
Gene Name
CD99 molecule like 2
Gene Aliases
CD99B, MIC2L1
Chromosome Location
Chr.X: 150766336 - 150898816 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184808.1 NP_001171737.1 2 - 3 360 83
NM_001242614.1 NP_001229543.1 2 - 3 360 83
NM_031462.3 NP_113650.2 2 - 3 360 83
XM_017029890.1 XP_016885379.1 2 - 3 224 83
GenBank mRNA AK023776.1 - 1 - 2 46 83
AK125020.1 - 2 - 3 171 83
AK290200.1 - 2 - 3 136 83
AK293244.1 - 2 - 3 166 83
AK295217.1 - 2 - 3 178 83
AL136580.1 - 2 - 3 250 83
AY078165.1 - 2 - 3 168 83
AY358837.1 - 2 - 3 233 83
BC030536.1 - 2 - 3 220 83
DC323066.1 - 2 - 3 360 83
KU178797.1 - 2 - 3 132 83

View Full Product Details