Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUGP1
Entrez Gene ID
57794
Gene Name
SURP and G-patch domain containing 1
Gene Aliases
F23858, RBP, SF4
Chromosome Location
Chr.19: 19276513 - 19320512 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011528155.2 XP_011526457.1 2 - 3 219 132
GenBank mRNA AK096152.1 - 2 - 3 219 132

View Full Product Details