The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012321.4 | NP_036453.1 | 1 - 2 | 176 | 73 |
GenBank mRNA | AF117235.1 | - | 1 - 2 | 139 | 73 |
AF182290.1 | - | 1 - 2 | 53 | 73 | |
AF251218.1 | - | 1 - 2 | 47 | 73 | |
BC000387.2 | - | 1 - 2 | 56 | 73 | |
BC003652.2 | - | 1 - 2 | 54 | 73 | |
BC022198.2 | - | 1 - 2 | 54 | 73 | |
BC023665.2 | - | 1 - 2 | 49 | 73 | |
DB493307.1 | - | 1 - 2 | 259 | 73 |