Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGL2
Entrez Gene ID
5863
Gene Name
ral guanine nucleotide dissociation stimulator like 2
Gene Aliases
HKE1.5, KE1.5, RAB2L
Chromosome Location
Chr.6: 33291654 - 33299388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243738.1 NP_001230667.1 11 - 12 1750 87
NM_004761.4 NP_004752.1 12 - 13 1947 87
XM_011514785.2 XP_011513087.1 10 - 11 1219 87
GenBank mRNA AB012295.1 - 11 - 12 1299 87
AK095967.1 - 9 - 10 921 87
AL050259.1 - 12 - 13 1520 87
BC000316.1 - 6 - 7 606 87
BC005037.1 - 9 - 10 988 87
BC032681.1 - 12 - 13 1502 87
BC040989.1 - 11 - 12 1802 87
BX648015.1 - 11 - 12 1292 87
U68142.1 - 6 - 7 553 87

View Full Product Details