The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004761.4 | NP_004752.1 | 1 - 2 | 519 | 100 |
GenBank mRNA | AL050259.1 | - | 1 - 2 | 92 | 100 |
BC032681.1 | - | 1 - 2 | 74 | 100 | |
BC040989.1 | - | 1 - 2 | 109 | 100 |