The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318876.1 | NP_001305805.1 | 6 - 7 | 744 | 64 |
NM_203290.3 | NP_976035.1 | 6 - 7 | 744 | 64 | |
XM_011515000.2 | XP_011513302.1 | 6 - 7 | 685 | 64 | |
GenBank mRNA | AF008442.1 | - | 6 - 7 | 670 | 64 |
AF047441.1 | - | 6 - 7 | 680 | 64 | |
AK293561.1 | - | 5 - 6 | 897 | 64 | |
BC008118.1 | - | 6 - 7 | 635 | 64 | |
BC008863.2 | - | 6 - 7 | 670 | 64 | |
BQ575535.1 | - | 3 - 4 | 64 |