The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318876.1 | NP_001305805.1 | 7 - 8 | 887 | 102 |
NM_203290.3 | NP_976035.1 | 7 - 8 | 887 | 102 | |
XM_011515000.2 | XP_011513302.1 | 7 - 8 | 828 | 102 | |
GenBank mRNA | AF008442.1 | - | 7 - 8 | 813 | 102 |
AF047441.1 | - | 7 - 8 | 823 | 102 | |
BC008118.1 | - | 7 - 8 | 778 | 102 | |
BC008863.2 | - | 7 - 8 | 813 | 102 | |
BQ575535.1 | - | 2 - 3 | 102 |