Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MED15
- Entrez Gene ID
-
51586
- Gene Name
- mediator complex subunit 15
- Gene Aliases
-
ARC105, CAG7A, CTG7A, PCQAP, TIG-1, TIG1, TNRC7
- Chromosome Location
-
Chr.22: 20507542 - 20587632 on Build GRCh38
View Full Product Details