The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282512.1 | NP_001269441.1 | 9 - 10 | 867 | 75 |
NM_006440.4 | NP_006431.2 | 9 - 10 | 867 | 75 | |
GenBank mRNA | AB019694.1 | - | 9 - 10 | 678 | 75 |
AB019695.1 | - | 9 - 10 | 772 | 75 | |
AB209724.1 | - | 9 - 10 | 645 | 75 | |
AF044212.1 | - | 9 - 10 | 688 | 75 | |
AF106697.1 | - | 9 - 10 | 867 | 75 | |
AF166126.1 | - | 4 - 5 | 752 | 75 | |
AF166127.1 | - | 9 - 10 | 772 | 75 | |
AF171054.1 | - | 9 - 10 | 685 | 75 | |
AF201385.1 | - | 9 - 10 | 675 | 75 | |
BC007489.2 | - | 8 - 9 | 562 | 75 | |
BC099923.1 | - | 9 - 10 | 679 | 75 | |
BC117354.1 | - | 9 - 10 | 647 | 75 | |
BX957216.1 | - | 9 - 10 | 669 | 75 | |
CR749428.1 | - | 9 - 10 | 669 | 75 |