The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160227.1 | NP_001153699.1 | 20 - 21 | 3557 | 73 |
NM_025137.3 | NP_079413.3 | 20 - 21 | 3557 | 73 | |
XM_006720700.1 | XP_006720763.1 | 20 - 21 | 3557 | 73 | |
XM_006720701.3 | XP_006720764.1 | 20 - 21 | 3557 | 73 | |
XM_017022634.1 | XP_016878123.1 | 20 - 21 | 3557 | 73 | |
XM_017022635.1 | XP_016878124.1 | 20 - 21 | 3557 | 73 | |
XM_017022636.1 | XP_016878125.1 | 9 - 10 | 1366 | 73 | |
GenBank mRNA | AB058743.2 | - | 20 - 21 | 3527 | 73 |
AB470308.1 | - | 20 - 21 | 3526 | 73 | |
AB470309.1 | - | 20 - 21 | 3526 | 73 | |
AB470310.1 | - | 20 - 21 | 3526 | 73 | |
AL834168.1 | - | 19 - 20 | 3278 | 73 | |
BC150640.1 | - | 20 - 21 | 3554 | 73 | |
BC153879.1 | - | 20 - 21 | 3527 | 73 |