The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160227.1 | NP_001153699.1 | 7 - 8 | 1633 | 100 |
NM_025137.3 | NP_079413.3 | 7 - 8 | 1633 | 100 | |
XM_006720700.1 | XP_006720763.1 | 7 - 8 | 1633 | 100 | |
XM_006720701.3 | XP_006720764.1 | 7 - 8 | 1633 | 100 | |
XM_017022634.1 | XP_016878123.1 | 7 - 8 | 1633 | 100 | |
XM_017022635.1 | XP_016878124.1 | 7 - 8 | 1633 | 100 | |
GenBank mRNA | AB058743.2 | - | 7 - 8 | 1603 | 100 |
AB470308.1 | - | 7 - 8 | 1602 | 100 | |
AB470309.1 | - | 7 - 8 | 1602 | 100 | |
AB470310.1 | - | 7 - 8 | 1602 | 100 | |
AL834168.1 | - | 6 - 7 | 1355 | 100 | |
BC047416.1 | - | 7 - 8 | 1613 | 100 | |
BC150640.1 | - | 7 - 8 | 1630 | 100 | |
BC153879.1 | - | 7 - 8 | 1603 | 100 |