Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPG11
Entrez Gene ID
80208
Gene Name
spastic paraplegia 11 (autosomal recessive)
Gene Aliases
ALS5, CMT2X, KIAA1840
Chromosome Location
Chr.15: 44562696 - 44663678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160227.1 NP_001153699.1 9 - 10 1917 128
NM_025137.3 NP_079413.3 9 - 10 1917 128
XM_006720700.1 XP_006720763.1 9 - 10 1917 128
XM_006720701.3 XP_006720764.1 9 - 10 1917 128
XM_017022634.1 XP_016878123.1 9 - 10 1917 128
XM_017022635.1 XP_016878124.1 9 - 10 1917 128
GenBank mRNA AB058743.2 - 9 - 10 1887 128
AB470308.1 - 9 - 10 1886 128
AB470309.1 - 9 - 10 1886 128
AB470310.1 - 9 - 10 1886 128
AL834168.1 - 8 - 9 1639 128
BC047416.1 - 9 - 10 1897 128
BC150640.1 - 9 - 10 1914 128
BC153879.1 - 9 - 10 1887 128

View Full Product Details