The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_126327.1 | 3 - 4 | 1086 | 72 | |
GenBank mRNA | BC009210.2 | - | 3 - 4 | 1036 | 72 |
BC012746.2 | - | 3 - 4 | 1036 | 72 |