The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201338.1 | NP_001188267.1 | 16 - 17 | 2459 | 126 |
NM_001201339.1 | NP_001188268.1 | 16 - 17 | 2456 | 126 | |
NM_001201340.1 | NP_001188269.1 | 15 - 16 | 2252 | 126 | |
NM_001320571.1 | NP_001307500.1 | 14 - 15 | 2184 | 126 | |
NM_001320572.1 | NP_001307501.1 | 16 - 17 | 2456 | 126 | |
NM_002967.3 | NP_002958.2 | 16 - 17 | 2459 | 126 | |
XM_017027114.1 | XP_016882603.1 | 14 - 15 | 2141 | 126 | |
XM_017027115.1 | XP_016882604.1 | 14 - 15 | 2141 | 126 | |
XM_017027116.1 | XP_016882605.1 | 15 - 16 | 1849 | 126 | |
XM_017027117.1 | XP_016882606.1 | 15 - 16 | 1846 | 126 | |
XM_017027118.1 | XP_016882607.1 | 15 - 16 | 1846 | 126 | |
GenBank mRNA | AK095736.1 | - | 12 - 13 | 1635 | 126 |
AK290444.1 | - | 16 - 17 | 2352 | 126 | |
AK293152.1 | - | 13 - 14 | 1957 | 126 | |
AK294671.1 | - | 14 - 15 | 1909 | 126 | |
AK295298.1 | - | 13 - 14 | 126 | ||
AK298707.1 | - | 15 - 16 | 2148 | 126 | |
AK299525.1 | - | 16 - 17 | 2217 | 126 | |
AK304474.1 | - | 12 - 13 | 1607 | 126 | |
BC126219.1 | - | 16 - 17 | 2330 | 126 | |
BC143937.1 | - | 16 - 17 | 2327 | 126 | |
BC143938.1 | - | 16 - 17 | 2327 | 126 | |
BC143939.1 | - | 16 - 17 | 2327 | 126 | |
L43631.1 | - | 16 - 17 | 2136 | 126 | |
U72355.1 | - | 16 - 17 | 2341 | 126 |