The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014629.3 | NP_055444.2 | 4 - 5 | 662 | 70 |
XM_011534767.2 | XP_011533069.1 | 3 - 4 | 524 | 70 | |
GenBank mRNA | AB002292.2 | - | 4 - 5 | 661 | 70 |
AL137508.1 | - | 1 - 2 | 266 | 70 |