Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- D2HGDH
- Entrez Gene ID
-
728294
- Gene Name
- D-2-hydroxyglutarate dehydrogenase
- Gene Aliases
-
D2HGD
- Chromosome Location
-
Chr.2: 241734579 - 241768816 on Build GRCh38
View Full Product Details