The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009566.2 | NP_001009566.1 | 14 - 15 | 2908 | 87 |
NM_001302883.1 | NP_001289812.1 | 13 - 14 | 2851 | 87 | |
NM_014944.4 | NP_055759.3 | 13 - 14 | 2878 | 87 | |
GenBank mRNA | AB020718.1 | - | 14 - 15 | 2908 | 87 |
AF438482.1 | - | 13 - 14 | 2844 | 87 | |
AK000605.1 | - | 9 - 10 | 1518 | 87 | |
AK001591.1 | - | 1 - 2 | 87 | ||
AK289798.1 | - | 13 - 14 | 2086 | 87 | |
AK304813.1 | - | 13 - 14 | 2093 | 87 | |
AY545556.1 | - | 3 - 4 | 516 | 87 | |
AY753301.1 | - | 13 - 14 | 2085 | 87 | |
BC033902.2 | - | 13 - 14 | 2336 | 87 |