The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009566.2 | NP_001009566.1 | 15 - 16 | 3078 | 64 |
NM_001302883.1 | NP_001289812.1 | 14 - 15 | 3021 | 64 | |
NM_014944.4 | NP_055759.3 | 14 - 15 | 3048 | 64 | |
GenBank mRNA | AB020718.1 | - | 15 - 16 | 3078 | 64 |
AF438482.1 | - | 14 - 15 | 3014 | 64 | |
AK001591.1 | - | 2 - 3 | 356 | 64 | |
AK289798.1 | - | 14 - 15 | 2256 | 64 | |
AK304813.1 | - | 14 - 15 | 2263 | 64 | |
AY753301.1 | - | 14 - 15 | 2255 | 64 | |
BC033902.2 | - | 14 - 15 | 2506 | 64 |