The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009566.2 | NP_001009566.1 | 5 - 6 | 1442 | 98 |
NM_001302883.1 | NP_001289812.1 | 5 - 6 | 1442 | 98 | |
NM_014944.4 | NP_055759.3 | 4 - 5 | 1412 | 98 | |
GenBank mRNA | AB020718.1 | - | 5 - 6 | 1442 | 98 |
AF438482.1 | - | 4 - 5 | 1378 | 98 | |
AK289798.1 | - | 4 - 5 | 620 | 98 | |
AK304813.1 | - | 5 - 6 | 684 | 98 | |
AY753301.1 | - | 4 - 5 | 619 | 98 | |
BC033902.2 | - | 4 - 5 | 870 | 98 |