The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006593.3 | NP_006584.1 | 2 - 3 | 1167 | 85 |
GenBank mRNA | AK226050.1 | - | 1 - 2 | 1503 | 85 |
AK299533.1 | - | 1 - 2 | 67 | 85 | |
AK312567.1 | - | 2 - 3 | 942 | 85 | |
BC029289.1 | - | 2 - 3 | 928 | 85 | |
BC104844.1 | - | 2 - 3 | 1095 | 85 | |
BC113418.1 | - | 2 - 3 | 1095 | 85 | |
U49250.1 | - | 2 - 3 | 874 | 85 |