The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006593.3 | NP_006584.1 | 4 - 5 | 1444 | 131 |
GenBank mRNA | AK297438.1 | - | 3 - 4 | 612 | 131 |
AK299533.1 | - | 3 - 4 | 344 | 131 | |
AK312567.1 | - | 4 - 5 | 1219 | 131 | |
AK315865.1 | - | 3 - 4 | 339 | 131 | |
AK316161.1 | - | 3 - 4 | 475 | 131 | |
BC029289.1 | - | 4 - 5 | 1205 | 131 | |
BC104844.1 | - | 4 - 5 | 1372 | 131 | |
BC113418.1 | - | 4 - 5 | 1372 | 131 | |
U49250.1 | - | 4 - 5 | 1151 | 131 |