The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285440.1 | NP_001272369.1 | 33 - 34 | 4428 | 65 |
XM_006714463.3 | XP_006714526.1 | 33 - 34 | 4443 | 65 | |
XM_011514005.2 | XP_011512307.1 | 34 - 35 | 4515 | 65 | |
GenBank mRNA | CR749280.1 | - | 33 - 34 | 4409 | 65 |