Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RICTOR
Entrez Gene ID
253260
Gene Name
RPTOR independent companion of MTOR complex 2
Gene Aliases
AVO3, PIA, hAVO3
Chromosome Location
Chr.5: 38937920 - 39074421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285439.1 NP_001272368.1 4 - 5 290 141
NM_001285440.1 NP_001272369.1 4 - 5 290 141
NM_152756.4 NP_689969.2 4 - 5 290 141
XM_006714463.3 XP_006714526.1 4 - 5 303 141
XM_011514005.2 XP_011512307.1 4 - 5 303 141
XM_017009311.1 XP_016864800.1 3 - 4 223 141
XM_017009312.1 XP_016864801.1 3 - 4 223 141
XM_017009313.1 XP_016864802.1 3 - 4 244 141
XM_017009314.1 XP_016864803.1 4 - 5 281 141
XM_017009315.1 XP_016864804.1 4 - 5 281 141
GenBank mRNA AK128358.1 - 3 - 4 223 141
AY515854.1 - 4 - 5 286 141
BC029608.1 - 4 - 5 281 141
BC137163.1 - 4 - 5 283 141
BC137164.1 - 4 - 5 283 141
BC144509.1 - 4 - 5 283 141
BP245264.1 - 4 - 5 290 141
CR749280.1 - 4 - 5 271 141

View Full Product Details