Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RICTOR
Entrez Gene ID
253260
Gene Name
RPTOR independent companion of MTOR complex 2
Gene Aliases
AVO3, PIA, hAVO3
Chromosome Location
Chr.5: 38937920 - 39074421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285439.1 NP_001272368.1 9 - 10 848 157
NM_001285440.1 NP_001272369.1 9 - 10 846 155
NM_152756.4 NP_689969.2 9 - 10 848 157
XM_006714463.3 XP_006714526.1 9 - 10 861 157
XM_011514005.2 XP_011512307.1 9 - 10 861 157
XM_011514006.2 XP_011512308.1 8 - 9 665 157
XM_017009311.1 XP_016864800.1 8 - 9 781 157
XM_017009312.1 XP_016864801.1 8 - 9 781 157
XM_017009313.1 XP_016864802.1 8 - 9 802 157
GenBank mRNA AK128358.1 - 8 - 9 781 157
AY515854.1 - 9 - 10 844 157
BC137163.1 - 9 - 10 841 157
BC137164.1 - 9 - 10 841 157
BC144509.1 - 9 - 10 841 157
CR749280.1 - 9 - 10 827 155

View Full Product Details