Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTN
Entrez Gene ID
7273
Gene Name
titin
Gene Aliases
CMD1G, CMH9, CMPD4, EOMFC, HMERF, LGMD2J, MYLK5, TMD
Chromosome Location
Chr.2: 178525989 - 178807423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256850.1 NP_001243779.1 41 - 42 9930 75
NM_001267550.2 NP_001254479.2 41 - 42 9930 75
NM_003319.4 NP_003310.4 40 - 41 9792 75
NM_133378.4 NP_596869.4 41 - 42 9930 75
NM_133379.4 NP_596870.2 41 - 42 9930 75
NM_133432.3 NP_597676.3 40 - 41 9792 75
NM_133437.4 NP_597681.4 40 - 41 9792 75
XM_017004819.1 XP_016860308.1 41 - 42 9933 75
XM_017004820.1 XP_016860309.1 41 - 42 9933 75
XM_017004821.1 XP_016860310.1 41 - 42 9930 75
XM_017004822.1 XP_016860311.1 41 - 42 9933 75
XM_017004823.1 XP_016860312.1 41 - 42 9933 75
GenBank mRNA X90568.1 - 40 - 41 9699 75

View Full Product Details