The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005895.3 | NP_005886.2 | 1 - 2 | 370 | 138 |
GenBank mRNA | AB027133.1 | - | 1 - 2 | 92 | 138 |
D63997.1 | - | 1 - 2 | 80 | 138 |