Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDI2
Entrez Gene ID
84301
Gene Name
DNA damage inducible 1 homolog 2
Gene Aliases
-
Chromosome Location
Chr.1: 15617458 - 15661057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032341.4 NP_115717.3 4 - 5 848 81
GenBank mRNA AK293006.1 - 4 - 5 848 81

View Full Product Details