Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC8
Entrez Gene ID
27181
Gene Name
sialic acid binding Ig like lectin 8
Gene Aliases
SAF2, SIGLEC-8, SIGLEC8L
Chromosome Location
Chr.19: 51450997 - 51458454 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011526735.2 XP_011525037.1 1 - 2 519 70
GenBank mRNA AF310234.1 - 1 - 2 464 70

View Full Product Details