Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBR5
Entrez Gene ID
51366
Gene Name
ubiquitin protein ligase E3 component n-recognin 5
Gene Aliases
DD5, EDD, EDD1, HYD
Chromosome Location
Chr.8: 102252273 - 102412719 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282873.1 NP_001269802.1 58 - 59 8641 65
NM_015902.5 NP_056986.2 58 - 59 8644 65
XM_005250962.4 XP_005251019.1 37 - 38 5411 65
XM_011517104.2 XP_011515406.2 57 - 58 8635 65
XM_011517105.2 XP_011515407.2 57 - 58 8632 65
XM_011517106.2 XP_011515408.2 57 - 58 8617 65
XM_017013533.1 XP_016869022.1 57 - 58 8629 65
XM_017013534.1 XP_016869023.1 57 - 58 8626 65
GenBank mRNA AB020703.3 - 58 - 59 8641 65
AF006010.2 - 58 - 59 8222 65
AK002172.1 - 11 - 12 1312 65
AK124995.1 - 1 - 2 929 65
AL833555.1 - 6 - 7 4662 65
BC044598.1 - 1 - 2 758 65
BC053948.1 - 1 - 2 758 65
BC137234.1 - 58 - 59 8216 65
U95000.1 - 58 - 59 8186 65

View Full Product Details