The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042681.1 | NP_001036146.1 | 20 - 21 | 4960 | 123 |
NM_001042682.1 | NP_001036147.1 | 10 - 11 | 3259 | 123 | |
NM_012102.3 | NP_036234.3 | 21 - 22 | 5145 | 123 | |
XM_005263464.2 | XP_005263521.1 | 20 - 21 | 4899 | 123 | |
XM_005263466.1 | XP_005263523.1 | 14 - 15 | 3782 | 123 | |
XM_011541510.1 | XP_011539812.1 | 19 - 20 | 4885 | 123 | |
XM_017001358.1 | XP_016856847.1 | 20 - 21 | 4650 | 123 | |
XM_017001359.1 | XP_016856848.1 | 20 - 21 | 5155 | 123 | |
GenBank mRNA | AB007927.2 | - | 19 - 20 | 4295 | 123 |
AB036737.1 | - | 21 - 22 | 4971 | 123 | |
AF016005.1 | - | 10 - 11 | 3237 | 123 | |
AF118275.1 | - | 6 - 7 | 2673 | 123 |