Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FANCE
Entrez Gene ID
2178
Gene Name
Fanconi anemia complementation group E
Gene Aliases
FACE, FAE
Chromosome Location
Chr.6: 35452339 - 35467106 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021922.2 NP_068741.1 3 - 4 1086 78
XM_005248885.2 XP_005248942.1 3 - 4 1108 78
XM_005248887.2 XP_005248944.1 3 - 4 1108 78
XM_005248888.3 XP_005248945.1 3 - 4 1108 78
XM_011514343.2 XP_011512645.1 3 - 4 801 78
GenBank mRNA AF265210.1 - 3 - 4 1086 78
AK292522.1 - 3 - 4 1097 78
AK312617.1 - 3 - 4 1074 78
BC046359.1 - 3 - 4 1092 78

View Full Product Details